Learn More
p73 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB7824
Description
Anti-p73 Antibody, α and β is a Rabbit Polyclonal Antibody for detection of p73 also known as p53-like Transcription Factor and has been validated in immunoprecipitation, western blot, immunohistochemistry (paraffin).
Specifications
p73 | |
Polyclonal | |
Purified immunoglobulin. 0.22 micron filtered. Provided at 1mg/mL in PBS, pH 7.4, containing 0.05% sodium azide. | |
Rabbit | |
100 μg | |
NM_005427.1 | |
2-8°C in undiluted aliquots for up to 12 months |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
TP73; p73; P73 | |
GST fusion protein containing aa450-636 of p73 protein. | |
Primary | |
Human | |