Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p75 LNGFR Mouse anti-Rat, Saporin, Clone: 192, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB390100UG
Description
Anti-p75 Low Affinity Nerve Growth Factor Receptor Antibody, Saporin conjugated, clone 192 is an antibody against p75 Low Affinity Nerve Growth Factor Receptor for use in 0.
Specifications
p75 LNGFR | |
192 | |
Saporin Conjugated. 192-IgG-SAP is sterile filtered and presented in Dulbecco's PBS with no preservative. | |
NGFR, p75(NTR), TNFRSF16, Gp80-LNGFR, CD271 | |
100 μg | |
Rat | |
IgG1 |
Monoclonal | |
Saporin | |
NM_002507.2 | |
Mouse | |
Primary | |
−70°C or below for long term. −30°C or 2-8°C for short term. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction