Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p75NTR (Neurotrophin Receptor) Rabbit anti-Human, Rat, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07476MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-476-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-476-MI Supplier MilliporeSigma Supplier No. 07476
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect p75NTR (Neurotrophin Receptor) using this Anti-p75NTR (Neurotrophin Receptor) Antibody validated for use in immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p75NTR (Neurotrophin Receptor)
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 70% storage buffer 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide and 30% glycerol.
Gene Symbols NGFR; Gp80-LNGFR; CD271; p75(NTR); TNFRSF16
Host Species Rabbit
Immunogen GST fusion protein corresponding to the intracellular domain (residues 274-425) of rat p75 neurotrophin receptor.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_005028.4
Target Species Human, Mouse, Rat
Content And Storage −20°C for up to 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.