Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p97/VCP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 8 publications
Supplier: Novus Biologicals NB1001558
Description
p97/VCP Polyclonal specifically detects p97/VCP in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
p97/VCP | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
15S Mg(2+)-ATPase p97 subunit, IBMPFDMGC8560, MGC131997, p97, TER ATPase, TERA, transitional endoplasmic reticulum ATPase, valosin containing protein, Valosin-containing proteinMGC148092, yeast Cdc48p homolog | |
Rabbit | |
89 kDa | |
0.1 mL | |
Cellular Markers, DNA Repair, Membrane Trafficking and Chaperones, Signal Transduction | |
7415 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
Western Blot 1:5000-1:15000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 1-4 ug/mg of lysate, Immunohistochemistry-Frozen | |
P55072 | |
VCP | |
The immunogen recognized by this antibody maps to a region between residue 750 and the C-terminus (residue 806) of human Valosin-Containing Protein using the numbering given in entry NP_009057.1 (GeneID 7415). | |
Affinity Purified | |
RUO | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Gorilla, Chimpanzee, Tasmanian devil, White-tufted-ear marmoset, Crab-eating macaque and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction