Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

proliferation-associated 2G4, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017922 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-922 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-922 Supplier Abnova Corporation Supplier No. H00005036D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human PA2G4 protein.

This gene encodes an RNA-binding protein that is involved in growth regulation. This protein is present in pre-ribosomal ribonucleoprotein complexes and may be involved in ribosome assembly and the regulation of intermediate and late steps of rRNA processing. This protein can interact with the cytoplasmic domain of the ErbB3 receptor and may contribute to transducing growth regulatory signals. This protein is also a transcriptional co-repressor of androgen receptor-regulated genes and other cell cycle regulatory genes through its interactions with histone deacetylases. This protein has been implicated in growth inhibition and the induction of differentiation of human cancer cells. Six pseudogenes, located on chromosomes 3, 6, 9, 18, 20 and X, have been identified. [provided by RefSeq

Sequence: MSGEDEQQEQTIAEDLVVTKYKMGGDIANRVLRSLVEASSSGVSVLSLCEKGDAMIMEETGKIFKKEKEMKKGIAFPTSISVNNCVCHFSPLKSDQDYILKEGDLVKIDLGVHVDGFIANVAHTFVVDVAQGTQVTGRKADVIKAAHLCAEAALRLVKPGNQNTQVTEAWNKVAHSFNCTPIEGMLSHQLKQHVIDGEKTIIQNPTDQQKKDHEKAEFEVHEVYAVDVLVSSGEGKAKDAGQRTTIYKRDPSKQYGLKMKTSRAFFSEVERRFDAMPFTLRAFEDEKKARMGVVECAKHELLQPFNVLYEKEGEFVAQFKFTVLLMPNGPMRITSGPFEPDLYKSEMEVQDAELKALLQSSASRKTQKKKKKKASKTAENATSGETLEENEAGD

Specifications

Antigen proliferation-associated 2G4, 38kDa
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human PA2G4 protein.
Formulation PBS with no preservative; pH 7.4
Gene PA2G4
Gene Accession No. BC007561
Gene Alias EBP1/HG4-1/p38-2G4
Gene Symbols PA2G4
Host Species Rabbit
Immunogen PA2G4 (NP_006182.2, 1 a.a. ∼ 394 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 5036
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.