Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PABP 1 Mouse anti-Human, Clone: 10E10, MilliporeSigma™

Catalog No. 041467MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-146-7MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-146-7MI Supplier MilliporeSigma Supplier No. 041467
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PABP 1, clone 10E10 is a Mouse Monoclonal Antibody for detection of PABP 1 also known as PABP 1, Poly(A)-binding protein 1, poly(A) binding protein cytoplasmic 1 and has been validated in western blot, immunocytochemistry and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PABP 1
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 10E10
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgGκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_002559.2
Gene Symbols PABPC1, PAB1, PABP, PABP1, PABPC2, PABPL1
Host Species Mouse
Immunogen Recombinant protein corresponding to human PABP 1.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.