Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PACSIN1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB10439MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10439MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10439MI Supplier MilliporeSigma Supplier No. AB10439
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-PACSIN1 Antibody, N-terminus detects level of PACSIN1 and has been published and validated for use in western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PACSIN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS containing 0.09% NaN3.
Gene Symbols KIAA1379; OTTHUMP00000016231; SDPI
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide selected from the N-terminal region of human PACSIN1.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) Q9BY11
Target Species Human, Mouse
Content And Storage 2-8°C for up to 6 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.