Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PADI4 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 09796MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-796-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-796-MI Supplier MilliporeSigma Supplier No. 09796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-PADI4 Antibody, C-terminus is an antibody against PADI4 for use in western blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PADI4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS and 0.09% sodium azide.
Gene Symbols PADI4; PAD; PAD4; PADI5; PDI4; PDI5
Host Species Rabbit
Immunogen KLH-conjugated synthetic linear peptide corresponding to the C-terminal region of PADI4.
Purification Method Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis against PBS.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_036519
Target Species Human
Content And Storage 2-8°C in undiluted aliquotes for up to 6 months, −20°C for long term. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.