Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PAI-1 Clone: MA-33H1F7,

Catalog No. 528217100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
52-821-7100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-821-7100UG Supplier MilliporeSigma™ Supplier No. 528217100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects PAI-1 Clone: MA-33H1F7 in Human, Mouse samples, and it is validated for Neutralization

TRUSTED_SUSTAINABILITY

Specifications

Antigen PAI-1
Applications Neutralization
Classification Monoclonal
Clone MA-33H1F7
Conjugate Unconjugated
Formulation In 100mM NaCl, 50mM sodium phosphate, pH 7.4.
Host Species Mouse
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽50kDa PAI-1 protein.
Target Species Human, Mouse
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.