Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAI-1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 09726MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-726-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-726-MI Supplier MilliporeSigma Supplier No. 09726

Rabbit Polyclonal Antibody

This Anti-PAI-1 Antibody is validated for use in western blotting for the detection of PAI-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAI-1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 100 μg of Protein A purified rabbit polyclonal IgG in 1X PBS with 0.1% sodium azide.
Gene Symbols SERPINE1; PAI; PLANH1; PAI-1; PAI1
Host Species Rabbit
Immunogen Recombinant human PAI-1
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000602.1
Target Species Human
Content And Storage −20°C for 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.