Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Paired box protein Pax-1 Clone: 7F11.2,

Catalog No. MABE978MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABE978MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE978MI Supplier MilliporeSigma™ Supplier No. MABE978
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Paired box protein Pax-1 Clone: 7F11.2 in Human samples, and it is validated for Western Blotting

Paired box protein Pax-1 (UniProt P15863; also known as HuP48) is encoded by the PAX1 (also known as HUP48, OFC2) gene (Gene ID 5075) in human. Pax-1 belongs to the family of transcription factors with conserved DNA-binding paired box domain. The family of paired box (PAX) genes are divided into subgroup I (PAX1 and PAX9), II (PAX2, PAX5, and PAX8), III (PAX3 and PAX7), and IV (PAX4 and PAX6). Pax1 & Pax9 are paralogous genes with overlapping expression and function. Pax1 is required for the proper development of the ventral structures of the vertebral column and double homozygous mutants of Pax1 and Pax9 result in a dramatic increase in the severity of the vertebral defects. During the early stages of chondrogenic differentiation, Pax1 induces and transactivates the transcriptional repressor Nk3 homeobox 2 (Nkx3.2) that promotes chondrogenesis of sclerotomal cells, while Pax1 act as a negative regulator during chondrogenic differentiation by antagonizing against Sox9-driven chondrocyte maturation. Pax1 is downregulated as cartilage formation proceeds and becomes restricted to the mesenchymal condensations that give rise to the anlagen of the intervertebral discs, and to the perichondrium surrounding the cartilaginous vertebral bodies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Paired box protein Pax-1
Applications Western Blot
Classification Monoclonal
Clone 7F11.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P15863
Gene Symbols PAX1, HUP48
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to the N-terminal half of human Paired box protein Pax-1.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_006183
Test Specificity Reactivity toward spliced isoforms 2 and 3 has not been determined.
Target Species Human
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.