Learn More
Paired mesoderm homeobox protein 2B (PHOX2B) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Paired mesoderm homeobox protein 2B (PHOX2B) |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal in buffer containing 0.5X PBS, 0.1 % BSA, 0.01% Thimerosal, 0.5% sodium azide, and 50% glycerol. |
| Gene Symbols | PHOX2B; PMX2B |
| Host Species | Rabbit |
| Immunogen | Linear peptide corresponding to human Paired mesoderm homeobox protein 2B (PHOX2B). |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.