Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAK6, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002995 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-995 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-995 Supplier Abnova Corporation Supplier No. H00056924D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human PAK6 protein.

This gene encodes a member of the p21-activated kinase (PAK) family. The proteins of this family are Rac/Cdc42-associated Ste20-like Ser/Thr protein kinases, characterized by a highly conserved amino-terminal Cdc42/Rac interactive binding (CRIB) domain and a carboxyl-terminal kinase domain. PAK kinases are implicated in the regulation of a number of cellular processes, including cytoskeleton rearrangement, apoptosis and the MAP kinase signaling pathway. The protein encoded by this gene was found to interact with androgen receptor (AR), which is a steroid hormone-dependent transcription factor that is important for male sexual differentiation and development. This gene was found to be highly expressed in testis and prostate tissues and the encoded protein was shown to cotranslocate into the nucleus with AR in response to androgen. Alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Sequence: MFRKKKKKRPEISAPQNFQHRVHTSFDPKEGKFVGLPPQWQNILDTLRRPKPVVDPSRITRVQLQPMKTVVRGSAMPVDGYISGLLNDIQKLSVISSNTLRGRSPTSRRRAQSLGLLGDEHWATDPDMYLQSPQSERTDPHGLYLSCNGGTPAGHKQMPWPEPQSPRVLPNGLAAKAQSLGPAEFQGASQRCLQLGACLQSSPPGASPPTGTNRHGMKAAKHGSEEARPQSCLVGSATGRPGGEGSPSPKTRESSLKRRLFRSMFLSTAATAPPSSSKPGPPPQSKPNSSFRPPQKDNPPSLVAKAQSLPSDQPVGTFSPLTTSDTSSPQKSLRTAPATGQLPGRSSPAGSPRTWHAQISTSNLYLPQDPTVAKGALAGEDTGVVTHEQFKAALRMVVDQGDPRLLLDSYVKIGEGSTGIVCLAREKHSGRQVAVKMMDLRKQQRRELLFNEVVIMRDYQHFNVVEMYKSYLVGEELWVLMEFLQGGALTDIVSQVRLNEEQIATVCEAVLQALAYLHAQGVIHRDIKSDSILLTLDGRVKLSDFGFCAQISKDVPKRKSLVGTPYWMAPEVISRSLYATEVDIWSLGIMVIEMVDGEPPYFSDSPVQAMKRLRDSPPPKLKNSHKVSPVLRDFLERMLVRDPQERATAQELLDHPFLLQTGLPECLVPLIQLYRKQTSTC

Specifications

Antigen PAK6
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human PAK6 protein.
Formulation PBS with no preservative; pH 7.4
Gene PAK6
Gene Accession No. NM_020168.3
Gene Alias PAK5
Gene Symbols PAK6
Host Species Rabbit
Immunogen PAK6 (NP_064553.1, 1 a.a. ∼ 681 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56924
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.