Learn More
p21 protein (Cdc42/Rac)-activated kinase 6, Mouse, MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human PAK6 protein.
Supplier: Abnova Corporation H00056924B01
Description
This gene encodes a member of the p21-activated kinase (PAK) family. The proteins of this family are Rac/Cdc42-associated Ste20-like Ser/Thr protein kinases, characterized by a highly conserved amino-terminal Cdc42/Rac interactive binding (CRIB) domain and a carboxyl-terminal kinase domain. PAK kinases are implicated in the regulation of a number of cellular processes, including cytoskeleton rearrangement, apoptosis and the MAP kinase signaling pathway. The protein encoded by this gene was found to interact with androgen receptor (AR), which is a steroid hormone-dependent transcription factor that is important for male sexual differentiation and development. This gene was found to be highly expressed in testis and prostate tissues and the encoded protein was shown to cotranslocate into the nucleus with AR in response to androgen. Alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Sequence: MFRKKKKKRPEISAPQNFQHRVHTSFDPKEGKFVGLPPQWQNILDTLRRPKPVVDPSRITRVQLQPMKTVVRGSAMPVDGYISGLLNDIQKLSVISSNTLRGRSPTSRRRAQSLGLLGDEHWATDPDMYLQSPQSERTDPHGLYLSCNGGTPAGHKQMPWPEPQSPRVLPNGLAAKAQSLGPAEFQGASQRCLQLGACLQSSPPGASPPTGTNRHGMKAAKHGSEEARPQSCLVGSATGRPGGEGSPSPKTRESSLKRRLFRSMFLSTAATAPPSSSKPGPPPQSKPNSSFRPPQKDNPPSLVAKAQSLPSDQPVGTFSPLTTSDTSSPQKSLRTAPATGQLPGRSSPAGSPRTWHAQISTSNLYLPQDPTVAKGALAGEDTGVVTHEQFKAALRMVVDQGDPRLLLDSYVKIGEGSTGIVCLAREKHSGRQVAVKMMDLRKQQRRELLFNEVVIMRDYQHFNVVEMYKSYLVGEELWVLMEFLQGGALTDIVSQVRLNEEQIATVCEAVLQALAYLHAQGVIHRDIKSDSILLTLDGRVKLSDFGFCAQISKDVPKRKSLVGTPYWMAPEVISRSLYATEVDIWSLGIMVIEMVDGEPPYFSDSPVQAMKRLRDSPPPKLKNSHKVSPVLRDFLERMLVRDPQERATAQELLDHPFLLQTGLPECLVPLIQLYRKQTSTCSpecifications
p21 protein (Cdc42/Rac)-activated kinase 6 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human PAK6 protein. | |
PAK6 | |
PAK5 | |
Mouse | |
50 μL | |
Kinases | |
56924 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
NM_020168.3 | |
PAK6 | |
PAK6 (NP_064553.1, 1 a.a. ∼ 681 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.