Learn More
PALB2 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABC331
Description
Anti-PALB2 Antibody is a rabbit polyclonal antibody, validated for use in western blotting and immunohistochemistry.
Specifications
PALB2 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
2-8°C for 12 months |
Immunohistochemistry, Western Blot | |
Unconjugated | |
PALB2 | |
KLH-conjugated linear peptide corresponding to human PALB2. | |
100 μg | |
Human |