Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pan Carcinoma, 40kDa Mouse anti-Human, Unlabeled, Clone: KS1/4, BD

Catalog No. BDB554276 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554276 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554276 Supplier BD Biosciences Supplier No. 554276
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KS1/4 antigen (Ag), defined by the KS1/4 antibody (KS1/4), is expressed in many epithelial-derived carcinomas and normal epithelial cell surfaces. The cDNA encoding KS1/4 has been isolated and contains an open reading frame of 314 amino acids including a putative signal sequence. KS1/4 Ag migrates as three glycosylated polypeptides with molecular masses of 35, 40 and 42kDa. The 40 and 42kDa species are similar proteins and appear to differ only by their degree of N-linked glycosylation. The 35kDa species results from proteolytic cleavages of the larger molecular weight proteins. KS1/4 was first described as a monoclonal antibody which recognized a lung adenocarcinoma associated antigen. In oncolytic drug targeting studies, KS1/4 suppressed the growth of human lung tumor xenografts in athymic nude mice. Subsequent studies showed that KS1/4 reacts with a variety of tumor tissues including colon, breast, ovarian, and pancreas. It also recognizes normal epithelium including colon, stomach, small intestine, liver, kidney, lung, pancreas, skin and ovary. These results suggest that KS1/4 Ag represents an epithelial cell/epithelial-derived carcinoma marker. KS1/4 recognizes a 40 - 42kDa antigen expressed on the cell surfaces of a
variety of epithelial tumors and normal epithelial cell types. KS1/4 also recognizes a 35kDa proteolytic fragment. UCLA-P3 cells derived from a human adenocarcinoma of the lung were used as immunogen.

Host Species: Mouse
Clone: KS1/4
Isotype: IgG2a
Species Reactivity: Human
Immunogen: UCLA-P3 cells

Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen Pan Carcinoma, 40kDa
Applications Flow Cytometry
Classification Monoclonal
Clone KS1/4
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen UCLA-P3 cells
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.