Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pan Histone Mouse anti-Human, FITC, Clone: H11-4, MilliporeSigma™

Catalog No. FCMAB390FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB390FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB390FMI Supplier MilliporeSigma Supplier No. FCMAB390F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Milli-Mark™ Anti-pan Histone-FITC Antibody, clone H11-4 is validated for use in FC for the detection of pan Histone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen pan Histone
Applications Flow Cytometry
Classification Monoclonal
Clone H11-4
Conjugate FITC
Formulation Purified mouse monoclonal IgG1 conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Host Species Mouse
Immunogen Purified nuclear fractions of HeLa cells
Purification Method Protein A Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from Light.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.