Learn More
Pan-MAGUK Mouse anti-Rat, Human, Mouse, Clone: K28/86, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma MABN72
Catalog No. MABN72MI
Only null left
Add to Cart
Description
Description
Anti-pan-MAGUK Antibody, clone K28/86 is an antibody against pan-MAGUK for use in immunohistochemistry, western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
pan-MAGUK
Monoclonal
Unconjugated
NP_001356
Mouse
Protein G
Primary
2-8°C for one year
IgG1 λ
Immunohistochemistry, Immunoprecipitation, Western Blot
K28/86
Purified mouse monoclonal IgG1λ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
DLG4, SAP90, PSD95, SAP-90, PSD-95
Recombinant protein corresponding to a fusion domain of human PSD95/SAP90 proteins.
100 μg
Human, Mouse, Rat
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only