Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuro-Chrom™ Pan Neuronal Marker Rabbit anti-Rat, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABN2300MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN2300MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN2300MI Supplier MilliporeSigma Supplier No. ABN2300
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Neuro-Chrom Pan Neuronal Marker-Rabbit is an antibody targeting the Pan Neuronal Marker protein, validated for use in immunocytochemistry, immunohistochemistry and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pan Neuronal Marker
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Blended polyclonal antibody cocktail in PBS with 0.05% NaN3
Host Species Rabbit
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage 2-8°C for up to 12 months
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.