Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Pan-NMDA NR1 Clone: R1JHL,

Catalog No. 45457815UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
15 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
45-457-815UG 15 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-457-815UG Supplier MilliporeSigma™ Supplier No. 45457815UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Pan-NMDA NR1 Clone: R1JHL in Rat samples, and it is validated for Immunoprecipitation

The rat NMDAR1 (NR1) was the first subunit of the NMDAR to be cloned. NR1 can form NMDA-activated channels when expressed in oocytes, but the currents are larger when combined with one or more of the NMDAR2 (NR2 A-D) subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pan-NMDA NR1
Applications Immunoprecipitation
Classification Monoclonal
Clone R1JHL
Conjugate Unconjugated
Formulation Lyophilized from cell culture supernatant.
Host Species Mouse
Immunogen a recombinant fusion protein consisting of amino acids 1-564 of rat NMDA NR1
Quantity 15 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽115kDa NMDA receptor NR1 subunit in rat cortex tissue extracts.
Target Species Rat
Content And Storage 2°C to 8°C
Form Ascites
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.