Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Pan-Ras Clone: F132-62,

Catalog No. OP21100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
OP21100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OP21100UG Supplier MilliporeSigma™ Supplier No. OP21100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Pan-Ras Clone: F132-62 in Human, Mouse, Rat samples, and it is validated for Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation

The human ras gene family consists of three identified members, H-, K- and N-ras, encoding proteins of 188-189 amino acids and 21,000 (p21) molecular weight. Human H- and K-ras are the homologues of v-H- and v-K-ras sequences originally isolated from Harvey and Kirsten strains of rat sarcoma viruses. Normal human cellular ras genes can be activated to oncogenes by mutations occurring in codons 12, 13 and 61; such mutated, activated and transforming ras genes have been identified and isolated from human tumors and cultured tumor cells. Although the expression patterns of ras proto-oncogene proteins in normal human tissues are known, similar information for activated ras oncogene encoded p21s and their relevance to human disease diagnosis and prognosis remains to be determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pan-Ras
Applications Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone F132-62
Conjugate Unconjugated
Formulation In 0.05M sodium phosphate buffer, 0.2% gelatin.
Host Species Mouse
Immunogen recombinant p21 Ras
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽21kDa H-, K-, and N-Ras proteins including both c- and v- Ras in SW480 and Y1 cells and normal skin tissue.
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.