Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pan-Shank Mouse anti-Rat, Mouse, Human, Clone: N23B/49, MilliporeSigma™

Catalog No. MABN24MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN24MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN24MI Supplier MilliporeSigma Supplier No. MABN24
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect pan-Shank using this Anti-pan-Shank Antibody, clone N23B/49 validated for use in western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen pan-Shank
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone N23B/49
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001004133
Gene Symbols SHANK2, CORTBP1, ProSAP1, CTTNBP1, SPANK-3, SHANK, PROSAP1, CortBP1, Shank2
Host Species Mouse
Immunogen Recombinant protein consisting of SH3/PDZ domain of rat Shank2.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.