Learn More
Panendothelial Cell Antigen Rat anti-Mouse, Unlabeled, Clone: MECA-32, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 553849
Description
The MECA-32 antibody reacts with a dimer of 50-55kDa subunits expressed on most or all endothelial cells in the embryonic and adult mouse, with the exception of cardiac and skeletal muscle and the brain. Normally in skeletal and cardiac muscle, MECA-32 an

Specifications
Panendothelial Cell Antigen | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse lymph node stromal cells | |
0.5 mg | |
Immunology | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
MECA-32 | |
Unconjugated | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.