Learn More
Panendothelial Cell Antigen Rat anti-Mouse, Unlabeled, Clone: MECA-32, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 550563
Description
The MECA-32 antibody reacts with a dimer of 50-55kDa subunits expressed on most or all endothelial cells in the embryonic and adult mouse, with the exception of cardiac and skeletal muscle and the brain. Normally in skeletal and cardiac muscle, MECA-32 an

Specifications
Panendothelial Cell Antigen | |
Monoclonal | |
31.25μg/mL | |
Plvap; Pv1; MECA32; Plasmalemma vesicle-associated protein | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
MECA-32 | |
Unconjugated | |
Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide. | |
Mouse lymph node stromal cells | |
1 mL | |
Immunology | |
Mouse | |
IgG2a κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.