Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pantothenate kinase 1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004641 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-641 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-641 Supplier Abnova Corporation Supplier No. H00053354A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PANK1.

This gene encodes a protein belonging to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. Alternative splicing has been observed at this locus and three variants, each encoding a distinct isoform, have been identified. [provided by RefSeq

Sequence: IRFPSCAMHRFIQMGSEKNFSSLHTTLCATGGGAFKFEEDFRMIADLQLHKLDELDCLIQGLLYVDSVGFNGKPECYYFENPTNPELCQKKPYCLDNPYP

Specifications

Antigen pantothenate kinase 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PANK1.
Formulation 50% glycerol
Gene PANK1
Gene Accession No. NM_148977
Gene Alias MGC24596/PANK/PANK1a/PANK1b
Gene Symbols PANK1
Host Species Mouse
Immunogen PANK1 (NP_683878, 310 a.a. ∼ 409 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 53354
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.