Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

panMunc13 Mouse, Unlabeled, Clone: 32, BD

Catalog No. BDB610998 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610998 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610998 Supplier BD Biosciences Supplier No. 610998

Mouse Monoclonal Antibody

Release of neurotransmitters at the synaptic junction is a primary mechanism of neuronal communication and is regulated by the synaptic vesicle (SV) cycle. Several components of the SV cycle were initially characterized in C. elegans. One of those, unc-13, is involved in normal presynaptic function. Mammalian homologues of unc-13 are Munc13-1, 13-2, and 13-3. These neuron-specific proteins lack N-terminal homology, but are highly conserved in their C-terminal regions. Here they contain a phorbal ester binding C1-domain and two C2-domains with homology to the PKC Ca2+;/phospholipid-binding domain. The most abundant isoform, Munc 13-1, is a presynaptic receptor with high affinities for phorbol ester and DAG. In response to phorbol ester, Munc13-1 associates with the plasma membrane and acts as a stimulation dependent enhancer of neurotransmitter release. In addition, Munc13-1 interacts with components of the SV cycle (syntaxin, SNAP25, synaptobrevin, Doc2) and with a brain specific isoform of β-spectrin, a protein that interacts with the actin cytoskeleton. Thus, it is thought that Munc13 proteins function in signaling pathways that regulate the neuronal exocytic machinery.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen panMunc13
Applications Western Blot
Classification Monoclonal
Clone 32
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat Munc13-1 aa. 621-834
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.