Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Pannexin-1/PANX1, Polyclonal

Catalog No. ABN242MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN242MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN242MI Supplier MilliporeSigma™ Supplier No. ABN242
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Pannexin-1/PANX1 Clone: in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry, Western Blotting

Pannexin-1 (UniProt Q96RD7; also known as Innexin) is encoded by the PANX1 (also known as MRS1, PX1) gene (ORF UNQ2529/PRO6028; Gene ID 24145) in human. Pannexin 1 is a high-conductance, voltage-gated channel protein widely expressed in many organs and tissues, including the sensory systems (eye, inner ear, taste buds, and the olfactory epithelium) and both neuronal and non-neuronal cell types. Three pannexin family genes (PANX1-3) have been identified in human and mouse genomes. Pannexin proteins share structural and functional similarities and play important roles in many physiological and pathological processes, such as ATP release, Ca2+ homeostasis, synaptic neurotransmitters release, apoptosis, and immunological response. Pannexin-1 deficiency causes caspase-3 activation and apoptosis induction of hair cells in cochlear spiral limbus (SLM), resulting in hearing loss among Panx1-/- mice. Panx1 is also expressed in the retina, lens and cornea of the eye, where it plays an important role in mediating the crosstalk between immune and neuronal cells. Pannexin-1 channel opening, frequently together with the activation of purinergic receptors by extracellular ATP, is reported to have a great implication in diverse pathological conditions, including ischemia, trauma, HIV infection, inflammation, migraine, and cancer cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pannexin-1/PANX1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q96RD7
Gene Symbols PANX1, MRS1, PX1
Host Species Rabbit
Immunogen GST-tagged recombinant human Pannexin-1/PANX1 C-terminal fragment.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_056183
Test Specificity Expected to react with both spliced isoforms reported by UniProt (Q96RD7-1, Q96RD7-2).
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C for one year
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.