Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Papillomavirus Mouse anti-Human, Clone: C1P5, MilliporeSigma™

Catalog No. MAB874MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB874MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB874MI Supplier MilliporeSigma Supplier No. MAB874
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Papillomavirus using this Anti-Papillomavirus Antibody, 16, 18 E6 protein, clone C1P5 validated for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Papillomavirus
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone C1P5
Conjugate Unconjugated
Formulation Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Host Species Mouse
Immunogen C1P5 was made against HPV type 18 E6 protein produced in bacteria.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.