Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Par-2 Mouse anti-Mouse, Human, Clone: SAM11, MilliporeSigma™

Catalog No. MABF243MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF243MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF243MI Supplier MilliporeSigma Supplier No. MABF243
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Par-2 Antibody, clone SAM11 is an antibody against Par-2 for use in western blotting, immunohistochemistry and flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Par-2
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone SAM11
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_005233
Gene Symbols F2RL1, GPR11, PAR2
Host Species Mouse
Immunogen Linear peptide corresponding to the N-terminus of human Par-2.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.