Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parainfluenza 2 Blend Mouse anti-Human, Clone: 231/9F, 233/9A, MilliporeSigma™

Catalog No. MAB8443MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8443MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8443MI Supplier MilliporeSigma Supplier No. MAB8443
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Parainfluenza 2 Blend Antibody, clones 231/9F, 233/9A detects level of Parainfluenza 2 Blend and has been published and validated for use in ELISA and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Parainfluenza 2 Blend
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 231/9F, 233/9A, 233/9A
Conjugate Unconjugated
Formulation Ascites fluid
Host Species Mouse
Immunogen Greer
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.