Learn More
Description
Specifications
Specifications
| Antigen | Parkin |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Rabbit serum. Lyophilized. Reconstitute with 50μL of sterile distilled water. Centrifuge to remove insoluble material. Contains no preservative. |
| Gene Symbols | PARK2; PDJ; PRKN; LPRS2; AR-JP; parkin; EC 6.3.2.- |
| Host Species | Rabbit |
| Immunogen | A 19 amino acid peptide (RILGEEQYNRYQQYGAEEC) corresponding to amino acids 305-323 of the human parkin molecule. An internal peptide sequence was chosen to avoid the possibility of cross reactivity with ubiquitin. |
| Quantity | 50 μL |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.