Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

poly(A)-specific ribonuclease (deadenylation nuclease), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004225 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-225 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-225 Supplier Abnova Corporation Supplier No. H00005073A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PARN.

The protein encoded by this gene is a 3'-exoribonuclease, with similarity to the RNase D family of 3'-exonucleases. It prefers poly(A) as the substrate, hence, efficiently degrades poly(A) tails of mRNAs. Exonucleolytic degradation of the poly(A) tail is often the first step in the decay of eukaryotic mRNAs. This protein is also involved in silencing of certain maternal mRNAs during oocyte maturation and early embryonic development, as well as in nonsense-mediated decay (NMD) of mRNAs that contain premature stop codons. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: AESYRIQTYAEYMGRKQEEKQIKRKWTEDSWKEADSKRLNPQCIPYTLQNHYYRNNSFTAPSTVGKRNLSPSQEEAGLEDGVSGEISDTELEQTDSCAE

Specifications

Antigen poly(A)-specific ribonuclease (deadenylation nuclease)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PARN.
Formulation 50% glycerol
Gene PARN
Gene Accession No. NM_002582
Gene Alias DAN
Gene Symbols PARN
Host Species Mouse
Immunogen PARN (NP_002573, 501 a.a. ∼ 599 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5073
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.