Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARN Mouse anti-Human, Clone: 7A7.1, MilliporeSigma™

Catalog No. 051238MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-123-8MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-123-8MI Supplier MilliporeSigma Supplier No. 051238

Mouse Monoclonal Antibody

Use Anti-PARN Antibody, clone 7A7.1 (Mouse Monoclonal Antibody) validated in western blot, immunocytochemistry to detect PARN also known as Poly(A)-specific ribonuclease PARN, Deadenylating nuclease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARN
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 7A7.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_002573
Gene Symbols PARN, DAN
Host Species Mouse
Immunogen Linear peptide corresponding to human PARN.
Purification Method Ion exchange
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.