Learn More
PARP (214/215) cleavage site Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | PARP (214/215) cleavage site |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Dulbecco's phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3 (+/- 0.1), 50% glycerol with 1.0mg/mL BSA (IgG, protease free) as a carrier. 0.05% sodium azide |
| Gene Symbols | PARP1; poly(ADP-ribosyl)transferase; PARP-1; PARP; NAD; pADPRT-1; ADPRT1; Poly; ADPRT; PPOL; EC 2.4.2.30 |
| Host Species | Rabbit |
| Immunogen | The antiserum was produced against a chemically synthesized peptide corresponding to the N-terminus of cleavage site (214/215) of human PARP. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.