Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP Mouse, Unlabeled, Clone: 42, BD

Catalog No. BDB611038 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611038 50 μg
BDB611039 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611038 Supplier BD Biosciences Supplier No. 611038
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Poly(ADP-ribose) polymerase (PARP) is a constitutively expressed, abundant nuclear protein. It has been referred to as “a molecular nick sensor” due to its recognition of, and catalytic activation by, single or double strand DNA breaks. The most critical ad extensively studied role of PARP is its participation in DNA base excision repair. Following binding to damaged DNA, PARP uses NAD+ to synthesize branched polymers of poly(ADP-ribose) on nuclear target proteins, including itself. Such modification of PARP increases its negative charge and results in loss of interactionwith DNA due to electrostatic repulsion. This opens the damaged DNA to DNA repair proteins. The poly(ADP-ribose) molecule is quickly degraded by poly (ADP-ribose) glycohydrolase that is found in association with PARP. PARP contains N-terminal DNA-binding domain (DBD), a central automodification domain that accepts poly (ADP-ribose), and a C-terminal catalytic domain. PARP is one of the earliest proteins targeted by caspase-3 during apoptosis. Although this protein is central to DNA repair, it has additional DNA-related functions that remain to be investigated.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen PARP
Applications Western Blot
Classification Monoclonal
Clone 42
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human PARP aa. 22-219
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.