Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP Mouse anti-Human, Unlabeled, Clone: 4C10-5, BD

Catalog No. BDB556494 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556494 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556494 Supplier BD Biosciences Supplier No. 556494
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PARP [Poly(ADP-ribose) polymerase] is a 113kDa nuclear chromatin-associated enzyme that catalyzes the transfer of ADP-ribose units from NAD+ to a variety of nuclear proteins including topoisomerases, histones, and PARP itself. The catalytic activity of PARP is increased in non-apoptotic cells following DNA damage, and PARP is thought to play an important role in mediating the normal cellular response to DNA damage. Additionally, PARP is a target of the caspase protease activity associated with apoptosis. During apoptosis, PARP is cleaved from a 113kDa intact form into smaller 89kDa and 24kDa fragments. This process separates the amino-terminal DNA-binding domain of the enzyme from the carboxy-terminal catalytic domain resulting in the loss of normal PARP function. Although the role of PARP in apoptosis remains to be elucidated, PARP cleavage is considered to be a marker of apoptosis. The 4C10-5 antibody recognizes both the intact 113kDa form and 89kDa fragment of PARP. The 4C10-5 antibody has been reported to recognize both native and denatured PARP. Purified human PARP was used as the immunogen and the antibody reported to react with an epitope located in the NAD binding domain. In dot blot assays, the antibody reacts with the native enzyme in the presence or absence of bound DNA as well as after synthesis of covalently linked poly (ADP-ribose). The 4C10-5 antibody is routinely tested by western blot analysis of untreated Jurkat T cells and Jurkat T cells induced to undergo apoptosis. This antibody is routinely tested by Western blot analysis and immunofluorescent imaging. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 4C10-5
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human PARP

Bioimaging, Dot Blotting, Flow Cytometry, Immunoprecipitation, Intracellular Staining, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen PARP
Applications Western Blot
Classification Monoclonal
Clone 4C10-5
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human PARP
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.