Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP Mouse anti-Human, Unlabeled, Clone: 7D3-6, BD

Catalog No. BDB551024 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551024 50 μg
BDB551025 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB551024 Supplier BD Biosciences Supplier No. 551024
Add to Cart
Add to Cart

Includes

Purified Mouse Anti-Human PARP, Jurkat Cell Lysate

Mouse Monoclonal Antibody

PARP [Poly(ADP-ribose) polymerase] is a 116kDa nuclear chromatin-associated enzyme that catalyzes the transfer of ADP-ribose units from NAD+ to a variety of nuclear proteins including topoisomerases, histones, and PARP itself. The catalytic activity of PARP is increased in nonapoptotic cells following DNA damage, and PARP is thought to play an important role in mediating the normal cellular response to DNA damage. Additionally, PARP is a target of the caspase protease activity associated with apoptosis. During apoptosis, PARP is cleaved from the 116kDa intact form into 85kDa and 25kDa fragments. This process separates the amino-terminal DNA-binding domain of the enzyme from the carboxy-terminal catalytic domain resulting in the loss of normal PARP function. Although the role of PARP in apoptosis remains to be elucidated, PARP cleavage is considered to be a marker of apoptosis.

Host Species: Mouse
Clone: 7D3-6
Isotype: IgG1
Species Reactivity: Human
Immunogen: Purified human PARP

Flow Cytometry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen PARP
Applications Western Blot
Classification Monoclonal
Clone 7D3-6
Concentration 0.25mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified human PARP
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.