Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Partitioning-defective 3 Rabbit anti-Human, Rat, Monkey, Mouse, Canine, Polyclonal, MilliporeSigma™

Catalog No. 07330MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-330-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-330-MI Supplier MilliporeSigma Supplier No. 07330
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Partitioning-defective 3 Antibody is an antibody against Partitioning-defective 3 and has been published in over 20 citations for use in IC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Partitioning-defective 3
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.014M phosphate buffer, pH 7.6, 0.175M NaCl, 0.07% sodium azide and 30% glycerol.
Gene Symbols PAR3; PARD3; ASIP; PHIP; SE2-5T2; PAR-3; SE2-5L16; PARD-3; PARD3A; PAR3A; PAR3alpha; PAR3-alpha; Bazooka; FLJ21015; Baz; SE2-5LT1
Host Species Rabbit
Immunogen GST fusion protein corresponding to residues 457-744 of mouse PAR-3 (accession number AY02657).
Purification Method Protein A chromatography
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_062565
Target Species Canine, Human, Monkey, Mouse, Rat
Content And Storage −20°C for up to 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.