Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parvalbumin Guinea Pig anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB15738MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15738MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15738MI Supplier MilliporeSigma Supplier No. AB15738

Guinea Pig Polyclonal Antibody

Detect Parvalbumin using this Anti-Parvalbumin Antibody validated for use in immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Parvalbumin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid.
Gene Symbols PVALB; parvalbumin; D22S749; MGC116759
Host Species Guinea Pig
Immunogen Synthetic peptide.
Purification Method Serum
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002854.2
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.