Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Parvalbumin Mouse anti-Bovine, Canine, Fish, Feline, Frog, Goat, Human, Mouse, Pig, Rat, Rabbit, Clone: PARV-19, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1572
Description
Anti-Parvalbumin Antibody is an antibody against Parvalbumin for use in immunocytochemistry, immunohistochemistry (paraffin) and western blotting.
Specifications
Parvalbumin | |
Monoclonal | |
Unconjugated | |
NM_002854.2 | |
Mouse | |
Unpurified | |
Primary | |
−20°C for up to 12 months. | |
IgG1 |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
PARV-19 | |
Ascites mouse monoclonal IgG1 liquid with 0.1% sodium azide. | |
PVALB, MGC116759, parvalbumin, D22S749 | |
Parvalbumin purified from frog muscle | |
100 μL | |
Bovine, Canine, Feline, Fish, Frog, Goat, Human, Mouse, Pig, Rabbit, Rat | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction