Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parvovirus B19 Mouse anti-Human, Clone: 521-5D, MilliporeSigma™

Catalog No. MAB8292MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8292MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8292MI Supplier MilliporeSigma Supplier No. MAB8292
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Parvovirus B19 Antibody, clone 521-5D is validated for use in ELISA, immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin) for the detection of Parvovirus B19.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Parvovirus B19
Applications ELISA, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone 521-5D
Conjugate Unconjugated
Formulation Purified immunoglobulin. In 0.02M PB and 0.25M NaCl pH=7.6 with 0.1% NaN3 as a preservative.
Host Species Mouse
Immunogen B-19 virus purified from an acute serum specimen from a child with aplastic crisis (Anderson, 1986).
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.