Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Parvovirus B19 Mouse anti-Human, Clone: R92F6, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB8293
Description
Anti-Parvovirus B19 Antibody, aa 328-344 of VP2 capsid protein, clone R92F6 is an antibody against Parvovirus B19 for use in ELISA, immunofluoresence, immunohistochemistry (paraffin) and western blotting.
Specifications
Parvovirus B19 | |
Monoclonal | |
Unconjugated | |
Mouse | |
Protein A purfied | |
Primary | |
2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
R92F6 | |
Liquid in 20mM Phosphate buffer, pH7.6, and 0.25M NaCl with 0.1% sodium azide as a preservative. | |
Native parvovirus B19 purified from human plasma. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction