Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAS domain containing serine/threonine kinase, Mouse, Clone: 6B7, Abnova™

Catalog No. 89001529 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-529 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-529 Supplier Abnova Corporation Supplier No. H00023178M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant PASK.

PAS domains regulate the function of many intracellular signaling pathways in response to both extrinsic and intrinsic stimuli. PASK is an evolutionarily conserved protein present in yeast, flies, and mammals.[supplied by OMIM

Sequence: MEDGGLTAFEEDQRCLSQSLPLPVSAEGPAAQTTAEPSRSFSSAHRHLSRRNGLSRLCQSRTALSEDRWSSYCLSSLAAQNICTSKLHCPAAPEHTDPSE

Specifications

Antigen PAS domain containing serine/threonine kinase
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6B7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant PASK.
Formulation ascites with no preservative
Gene PASK
Gene Accession No. BC063585
Gene Alias DKFZp434O051/DKFZp686P2031/KIAA0135/PASKIN/STK37
Gene Symbols PASK
Host Species Mouse
Immunogen PASK (AAH63585, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 23178
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.