Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PATE1 Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABC244MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC244MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC244MI Supplier MilliporeSigma Supplier No. ABC244
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-PATE1 Antibody is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of PATE1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PATE1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols PATE1; PATE
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to human PATE1.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_612151
Target Species Human, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.