Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX-5 Rat, Alexa Fluor 647, Clone: 1H9, BD

Catalog No. BDB562814 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562814 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562814 Supplier BD Biosciences Supplier No. 562814

Rat Monoclonal Antibody

The 1H9 monoclonal antibody clone specifically binds to human and mouse Paired box protein Pax-5. Pax-5 is a ∽50 kDa protein that is also known as B-cell-specific transcription factor and B cell specific activator protein (BSAP). Pax-5 is a member of the paired box (Pax) family of transcription factors. It is expressed in pro-B, pre-B and mature B cells. Through the Groucho family of co-repressors, Pax-5 likely functions as a transcriptional repressor of non-B-lymphoid genes during the B cell commitment process. In the early stages of B cell development, Pax-5 influences the expression of several B-cell-specific genes, such as CD19 and CD20 and maintains B cell identity. Pax-5 suppression is involved in the upregulation of Blimp-1 leading to the development of Pax-5-negative plasma cells. Pax-5 mRNA is transiently detected in the mesencephalon and spinal cord during embryogenesis. Expression then shifts to the fetal liver and correlates with the onset of B lymphopoiesis. Altered forms and expression patterns of Pax-5 have been associated with some lymphoid and nonlymphoid cancers.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PAX-5
Applications Flow Cytometry
Classification Monoclonal
Clone 1H9
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Description Pax5, PAX5, KLP, BSAP, EBB-1; B-cell-specific transcription factor
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Recombinant Mouse Pax-5 protein containing aa 154-284
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.