Learn More
Description
Host Species: Rat
Clone 1H9
Isotype IgG2a, κ
Species Reactivity Hu, Ms
Immunogen Recombinant Mouse Pax-5 protein containing aa 154-284
Flow Cytometry, Intracellular Staining
Specifications
Specifications
| Antigen | PAX-5 |
| Classification | Monoclonal |
| Clone | 1H9 |
| Conjugate | Brilliant Violet 421 |
| Host Species | Rat |
| Immunogen | Recombinant Mouse Pax-5 protein containing aa 154-284 |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.