Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pax-5 Mouse, Unlabeled, Clone: 24, BD

Catalog No. BDB610863 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610863 150 μg
BDB610862 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610863 Supplier BD Biosciences Supplier No. 610863
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

There are at least nine members of the paired box (Pax) gene family whose protein products are transcription factors involved in development. The conserved paired box DNA-binding domain is found in the N-terminal region of Pax proteins. An octomer and homeodomain sequence are conserved in the center of the proteins. The Ser/Thr/Pro-rich region in the C-terminal portion contains a conserved 100 amino acid transactivating domain. One of the best studied Pax family members, Pax 5, is a B cell specific activator protein (BSAP). In the early stages of B cell development, Pax-5 influences the expression of several B-cell-specific genes, such as CD19 and CD20. Pax-5 is expressed primarily in pro-, pre-, and mature B cells, but not in plasma cells. Interestingly, Pax-5 mRNA is transiently detected in the mesencephalon and spinal cord during embryogenesis. Expression then shifts to the fetal liver and correlates with the onset of B lymphopoiesis. Pax-5 has been found to be important in both B cell and nervous system development.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Pax-5
Applications Western Blot
Classification Monoclonal
Clone 24
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Pax-5 aa. 151-306
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.