Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX3/7BP Rabbit anti-Human, Rat, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABD85MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABD85MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABD85MI Supplier MilliporeSigma Supplier No. ABD85
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-PAX3/7BP Antibody is an antibody against PAX3/7BP for use in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAX3/7BP
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Unpurified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.
Gene Symbols Paxbp1
Host Species Rabbit
Immunogen Recombinant protein corresponding to mouse PAX3/7BP.
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.