Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAX3 Mouse anti-Human, Mouse, Rat, Clone: 7D8G7, MilliporeSigma™

Catalog No. MABC699MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC699MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC699MI Supplier MilliporeSigma Supplier No. MABC699
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect PAX3 using this Anti-PAX3, clone 7D8G7 Antibody validated for use in western blotting and ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pax3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7D8G7
Conjugate Unconjugated
Formulation Mouse monoclonal IgG1 ascitic fluid with up to 0.1% sodium azide.
Gene Symbols PAX3, HUP2
Host Species Mouse
Immunogen Purified recombinant fragment of human PAX3 expressed in E. Coli
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.