Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

paired box 4, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017929 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-929 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-929 Supplier Abnova Corporation Supplier No. H00005078D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human PAX4 protein.

This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The paired box 4 gene is involved in pancreatic islet development and mouse studies have demonstrated a role for this gene in differentiation of insulin-producing beta cells. [provided by RefSeq

Sequence: MNQLGGLFVNGRPLPLDTRQQIVRLAVSGMRPCDISRILKVSNGCVSKILGRYYRTGVLEPKGIGGSKPRLATPPVVARIAQLKGECPALFAWEIQRQLCAEGLCTQDKTPSVSSINRVLRALQEDQGLPCTRLRSPAVLAPAVLTPHSGSETPRGTHPGTGHRNRTIFSPSQAEALEKEFQRGQYPDSVARGKLATATSLPEDTVRVWFSNRRAKWRRQEKLKWEMQLPGASQGLTVPRVAPGIISAQQSPGSVPTAALPALEPLGPSCYQLCWATAPERCLSDTPPKACLKPCWGHLPPQPNSLDSGLLCLPCPSSHCPLASLSGSQALLWPGCPLLYGLE

Specifications

Antigen paired box 4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human PAX4 protein.
Formulation PBS with no preservative; pH 7.4
Gene PAX4
Gene Accession No. NM_006193.1
Gene Alias KPD/MGC129960/MODY9
Gene Symbols PAX4
Host Species Rabbit
Immunogen PAX4 (NP_006184.1, 1 a.a. ∼ 343 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 5078
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.